Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBK Mouse, Unlabeled, Clone: 31, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612170

 View more versions of this product

Catalog No. BDB612170


Add to cart

Description

Description

Mitogen activated protein (MAP) kinase signal transduction pathways mediate the effects of various extracellular stimuli on biological processes such as proliferation, differentiation, and death. The p38 MAP kinases are activated by dual phosphorylation on Thr and Tyr within the motif Thr-Gly-Tyr located in kinase subdomain VIII. Activation of p38 MAPK is mediated specifically by the MAP kinase kinases, MKK3 and MKK6. PBK is a PDZ-binding Ser-Thr kinase related to the MKK3/6 MAPKK family. PBK was also identified as T-LAK celloriginated protein kinase (TOPK) and spermatogenesis-related protein kinase (SPK). PBK contains the conserved dual specficity active site sequence (D-X-K-X-X-N) at amino acids 174 to 179, and a C terminal ETDV motif that binds PDZ domains. The mRNA expression of PBK is reportedly highest in placenta, and lower in heart and pancreas. In mitotic cells, PBK is phosphorylated and enzymatically active, and in vitro, PBK is phosphorylated by Cdc2/cyclin B. PBK can bind to the PDZ domain-containing protein, Dlg, and can phosphorylate p38 MAPK. Thus, PBK is a MAPKK family member that may be important for regulation of PDZ domain proteins during mitosis.

Western Blotting

Specifications

Specifications

PBK
31
Unconjugated
Affinity Purified
Human PBK aa. 191-322
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.