Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBK Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513968

Catalog No. PIPA513968

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

PBK is a serine/threonine kinase related to the dual specific mitogen-activated protein kinase kinase (MAPKK) family. Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.
Specifications

Specifications

PBK
Polyclonal
PBS with 0.09% sodium azide
Q96KB5
T-lymphokine-activated killer cell-originated protein kinase; T-LAK cell-originated protein kinase; PDZ-binding kinase; Spermatogenesis-related protein kinase; SPK; MAPKK-like protein kinase; Nori-3; PBK; TOPK
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55872
Immunohistochemistry, Western Blot
Unconjugated
PBK
Liquid
PBK
KLH conjugated synthetic peptide between 286-317 amino acids from the C-terminal region of human PBK
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit