Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEBP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515363

Catalog No. PIPA515363

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PBP binds ATP, opioids and phosphatidylethanolamine, exhibiting a lower affinity for phosphatidylinositol and phosphatidylcholine. This serine protease inhibitor inhibits thrombin, neuropsin and chymotrypsin but not trypsin, tissue type plasminogen activator and elastase. PBP contains hippocampal cholinergic neurostimulating peptide (HCNP), which may be involved in the function of the presynaptic cholinergic neurons of the central nervous system. HCNP increases the production of choline acetyltransferase but not acetylcholinesterase.
Specifications

Specifications

PEBP1
Polyclonal
PBS with 0.09% sodium azide
P30086, P70296
Phosphatidylethanolamine-binding protein, PEBP, Prostatic binding protein, HCNPpp, Neuropolypeptide h3, Raf kinase inhibitor protein, RKIP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23980, 5037
Immunohistochemistry, Western Blot
Unconjugated
PEBP1
Liquid
PEBP1
KLH conjugated synthetic peptide between 137-167 amino acids from the central region of human PBP
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit