Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pre-B-cell leukemia homeobox 3, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant PBX3.

Manufacturer:  Abnova Corporation H00005090A01

Catalog No. 89-004-226


Add to cart

Description

Description

Sequence: FNLPNSGDMFMNMQSLNGDSYQGSQVGANVQSQVDTLRHVINQTGGYSDGLGGNSLYSPHNLNANGGWQDATTPSSVTSPTEGPGSVHSDTSN
Specifications

Specifications

pre-B-cell leukemia homeobox 3
Unconjugated
50% glycerol
NM_006195
PBX3
PBX3 (NP_006186, 342 a.a. ~ 434 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5090
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant PBX3.
PBX3
Serum
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit