Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PC1/PC3, Polyclonal, ™ Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ AB10553/DEL

Catalog No. 50-172-979

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein is a type I proinsulin-processing enzyme that plays a key role in regulating insulin biosynthesis. It is also known to cleave proopiomelanocortin, prorenin, proenkephalin, prodynorphin, prosomatostatin and progastrin. Mutations in this gene are thought to cause obesity. This encoded protein is associated with carcinoid tumors. The use of alternate polyadenylation sites has been found for this gene. Multiple alternatively spliced transcript variants have been described for this gene but their full length nature is not known.

Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Immunogen: This antibody is generated from rabbits immunized with a KLH conjugated linear peptide within human Prohormone Convertase PC1/PC3.

Immunocytochemistry, Immunohistochemistry, Western Blotting

Specifications

Specifications

PC1/PC3
Affinity Purified
This antibody is generated from rabbits immunized with a KLH conjugated linear peptide within human Prohormone Convertase PC1/PC3.
Primary
Human, Murine, Rat
Unconjugated
Rabbit
100μg
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit