Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPCAT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526318

Catalog No. PIPA526318


Add to cart

Description

Description

PCAT1 acyltransferase (LPCAT; EC 2.3.1.23) catalyzes the conversion of LPC to hosphatidylcholine (PC) in the remodeling pathway of PC biosynthesis.
Specifications

Specifications

LPCAT1
Polyclonal
PBS with 0.09% sodium azide
Q8NF37
AYTL2, PFAAP3, lpcat, 1-acylglycerophosphocholine O-acyltransferase, 1-alkylglycerophosphocholine O-acetyltransferase, LPC acyltransferase 1, LPCAT-1, acetyl-CoA:lyso-PAF acetyltransferase, acetyl-CoA:lyso-platelet-activating factor acetyltransferase, acyl-CoA:lysophosphatidylcholin
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79888
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
LPCAT1
Liquid
LPCAT1
KLH conjugated synthetic peptide between 506-534 amino acids from the C-terminal region of human PCAT1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit