Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCBD1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526121

Catalog No. PIPA526121

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

PCBD1 encodes pterin-4 alpha-carbinolamine dehydratase, an enzyme involved in phenylalanine hydroxylation. A deficiency of this enzyme leads to hyperphenylalaninemia. The enzyme regulates the homodimerization of the transcription factor hepatocyte nuclear factor 1 (HNF1).
Specifications

Specifications

PCBD1
Polyclonal
PBS with 0.09% sodium azide
P61457, P61458
DCOH, PCBD, PCD, PHS, 4-alpha-hydroxy-tetrahydropterin dehydratase, 6-pyruvoyl-tetrahydropterin synthase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1), dimerization cofactor of HNF1, dimerizing cofactor for HNF1, phenylalanine hydroxylase-stimulating protein, pt
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
13180, 5092
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PCBD1
Liquid
PCBD1
KLH conjugated synthetic peptide between 11-37 amino acids from the central region of human PCBD1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit