Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PCCA, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232215

Catalog No. NBP232215


Add to cart

Description

Description

PCCA Polyclonal antibody specifically detects PCCA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative).
Specifications

Specifications

PCCA
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunohistochemistry
NP_000273.2
EC 6.4.1, EC 6.4.1.3, pccA complementation group, PCCase alpha subunit, PCCase subunit alpha, propanoyl-CoA:carbon dioxide ligase alpha subunit, Propanoyl-CoA:carbon dioxide ligase subunit alpha, propionyl CoA carboxylase, alpha polypeptide, propionyl Coenzyme A carboxylase, alpha polypeptide, propionyl-CoA carboxylase alpha chain, mitochondrial
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Domestic duck, White-tufted-ear marmoset, Domestic pigeon,Three-spined stickleback and Southern platyfish.
Western Blot, Immunohistochemistry
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
PCCA
The immunogen this antibody was made to, maps to a region between residue 678 to 728 of human Propionyl CoA Carboxylase, Alpha Polypeptide using the numbering given in entry NP_000273.2 (GeneID 5095).
Immunogen affinity purified
RUO
Primary
5095
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only