Learn More
protocadherin alpha 6, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human PCDHA6 protein.
Supplier: Abnova Corporation H00056142D01P
Description
This gene is a member of the protocadherin alpha gene cluster, one of three related gene clusters tandemly linked on chromosome five that demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The alpha gene cluster is composed of 15 cadherin superfamily genes related to the mouse CNR genes and consists of 13 highly similar and 2 more distantly related coding sequences. The tandem array of 15 N-terminal exons, or variable exons, are followed by downstream C-terminal exons, or constant exons, which are shared by all genes in the cluster. The large, uninterrupted N-terminal exons each encode six cadherin ectodomains while the C-terminal exons encode the cytoplasmic domain. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins that most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been observed and additional variants have been suggested but their full-length nature has yet to be determined. [provided by RefSeq
Sequence: MVFTPEDRLGKQCLLLPLLLLAAWKVGSGQLHYSVPEEAKHGTFVGRIAQDLGLELAELVPRLFRMASKDREDLLEVNLQNGILFVNSRIDREELCGRSAECSIHLEVIVDRPLQVFHVDVEVRDINDNPPLFPVEEQRVLIYESRLPDSVFPLEGASDADVGSNSILTYKLSSSEYFGLDVKINSDDNKQIGLLLKKSLDREEAPAHNLFLTATDGGKPELTGTVQLLVTVLDVNDNAPTFEQSEYEVRIFENADNGTTVIRLNASDRDEGANGAISYSFNSLVAAMVIDHFSIDRNTGEIVIRGNLDFEQENLYKILIDATDKGHPPMAGHCTVLVRILDKNDNVPEIALTSLSLPVREDAQFGTVIALISVNDLDSGANGQVNCSLTPHVPFKLVSTFKNYYSLVLDSALDRESVSAYELVVTARDGGSPSLWATASLSVEVADMNDNAPAFAQPEYTVFVKENNPPGCHIFTVSARDADAQENALVSYSLVERRVGERALSSYISVHAESGKVYALQPLDHEELELLQFQVSARDAGVPPLGSNVTLQVFVLDENDNAPALLAPRVGGTGGAVSELVPRSLGAGQVVAKVRAVDADSGYNAWLSYELQPPASSARFPFRVGLYTGEISTTRVLDEADSPRHRLLVLVKDHGEPALTATATVLVSLVESGQAPKASSRASVGAAGPEAALVDVNVYLIIAICAVSSLLVLTLLLYTALRCSAPPTEGACTADKPTLVCSSAVGSWSYSQQRRQRVCSGEGPPKMDLMAFSPSLSPCPIMMGKAENQDLNEDHDAKPRQPNPDWRYSASLRAGMHSSVHLEEAGILRAGPGGPDQQWPTVSSATPEPEAGEVSPPVGAGVNSNSWTFKYGPGNPKQSGPGELPDKFIIPGSPAIISIRQEPTNSQIDKSDFITFGKKEETKKKKKKKKGNKTQEKKEKGNSTTDNSDQSpecifications
protocadherin alpha 6 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human PCDHA6 protein. | |
PCDHA6 | |
CNR2/CNRN2/CNRS2/CRNR2/PCDH-ALPHA6 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_018909.2 | |
PCDHA6 | |
PCDHA6 (NP_061732.1, 1 a.a. ∼ 950 a.a) full-length human protein. | |
100 μg | |
Cell Adhesion | |
56142 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.