Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHB11 Mouse anti-Human, Clone: 4F11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA522254

Catalog No. PIMA522254


Add to cart

Description

Description

Peptide Sequence: GSETWSFSVA EEMQSGSFVG NLAKDLGLKV RELSSRGARV VSNDKKQRLQ LDINTGDLLL SETLDREELC GSIEPCVLHL QVLMQNPTQF LQIELQVRDI

This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3 ft. exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections.
Specifications

Specifications

PCDHB11
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9Y5F2
ME2, PCDH-BETA11, PCDH-beta-11, cadherin ME2, protocadherin beta-11
Mouse
IgG2b, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
56125
ELISA
4F11
Unconjugated
PCDHB11
Liquid
PCDHB11
PCDHB11 (NP_061754, 27 a.a. ∽ 126 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit