Learn More
protocadherin gamma subfamily B, 1 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PCDHGB1 protein.
Supplier: Abnova Corporation H00056104B01
Description
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq
Sequence: MQRAREAEMMKSQVLFPFLLSLFCGAISQQIRYTIPEELANGSRVGKLAKDLGLSVRELPTRKLRVSAEDYFNVSLESGDLLVNGRIDREKICGRKLECALEFETVAENPMNVFHVVVVIQDINDNAPRFVAKGIDLEICESALPGVKFSLDSAQDADVEGNSLKLYTINPNQYFSLSTKESPDGSKYPVLLLEKPLDREHQSSHRLILTAMDGGDPPLSGTTHIWIRVTDANDNAPVFSQEVYRVSLQENVPWGTSVLRVMATDQDEGINAEITYAFLNSPISTSLFNLNPNTGDITTNGTLDFEETSRYVLSVEAKDGGVHTAHCNVQIEIVDENDNAPEVTFMSFSNQIPEDSDLGTVIALIKVRDKDSGQNGMVTCYTQEEVPFKLESTSKNYYKLVIAGALNREQTADYNVTIIATDKGKPALSSRTSITLHISDINDNAPVFHQASYVVHVSENNPPGASIAQVSASDPDLGPNGRVSYSILASDLEPRELLSYVSVSPQSGVVFAQRAFDHEQLRAFELTLQARDQGSPALSANVSLRVLVGDLNDNAPRVLYPALGPDGSALFDMVPRAAEPGYLVTKVVAVDADSGHNAWLSYHVLQASEPGLFSLGLRTGEVRTARALGDRDAARQRLLVAVRDGGQPPLSATATLHLIFADSLQEVLPDLSDRPEPSDPQTELQFYLVVALALISVLFLLAVILAIALRLRRSSSLDTEGCFQTGLCSKSGPGVPPNHSEGTLPYSYNLCIASHSAKTEFNSLNLTPEMAPPQDLLCDDPSMVVCASNEDHKIAYDPSLSSHVSFCKSSSpecifications
protocadherin gamma subfamily B, 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human PCDHGB1 protein. | |
PCDHGB1 | |
MGC119466/MGC119467/MGC119469/PCDH-GAMMA-B1 | |
Mouse | |
50 μL | |
Cell Adhesion | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_032095.1 | |
PCDHGB1 | |
PCDHGB1 (NP_115266.1, 1 a.a. ∼ 810 a.a) full-length human protein. | |
RUO | |
Yes | |
56104 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.