Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PCDHGB4, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP215065

Catalog No. NBP215065


Add to cart

Description

Description

PCDHGB4 Polyclonal specifically detects PCDHGB4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PCDHGB4
1mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Q9UN71
cadherin 20, Cadherin-20, CDH20cadherin-20, FIB2fibroblast cadherin FIB2, Fibroblast cadherin-2, MGC138293, MGC142167, PCDH-gamma-B4, protocadherin gamma subfamily B, 4, protocadherin gamma-B4
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
8641
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Affinity Purified
PCDHGB4
Recombinant protein encompassing a sequence within the N-terminus region of human PCDHGB4. The exact sequence is proprietary.
100 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human. Predicted cross-reactivity based on sequence identity: Chimpanzee (100%).
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only