Learn More
PCDHGB6, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human PCDHGB6 peptide using ARM Technology.
Supplier: Abnova Corporation H00056100K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
PCDHGB6 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
PCDHGB6 | |
PCDHGB6 | |
A synthetic peptide of human PCDHGB6 | |
RUO | |
56100 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
protocadherin gamma subfamily B, 6 | |
PCDHGB6 | |
PCDH-GAMMA-B6 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.