Learn More
PCDHGC5 Mouse anti-Human, Clone: 3A6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522245
Catalog No. PIMA522245
Add to cart
Description
Description
Peptide Sequence: RPPGSLLCTV AASDPDTGDN ARLTYSIVGN QVQGAPASSF VYVNPEDGRI FAQRTFDYEL LQMLQIVVGV RDSGSPPLHA NTSLHVFVLD ENDNAPAVLH PRPDWEHSA
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Specifications
Specifications
PCDHGC5
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q9Y5F6
PCDH-GAMMA-C5, protocadherin gamma-C5
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
56097
ELISA, Western Blot
3A6
Unconjugated
PCDHGC5
Liquid
PCDHGC5
PCDHGC5 (NP_061752, 467 a.a. ∽ 575 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human