Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Rabbit anti-Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Monkey, Xenopus, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178105

Catalog No. NBP178105


Add to cart

Description

Description

PCNA Polyclonal antibody specifically detects PCNA in Human, Mouse, Rat, Bovine, Canine, Chicken, Fish, Monkey, Xenopus samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PCNA
0.97 mg/ml
Western Blot 1:500-1:2000, ELISA 1:2000-1:8000
P12004
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Rabbit
IgG
0.1 mg
Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Stem Cell Markers
Primary
5111
Bovine, Canine, Chicken, Fish, Human, Monkey, Mouse, Rat, Xenopus
ELISA, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
PCNA
This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human PCNA protein.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
A BLAST analysis was used to suggest cross reactivity with PCNA protein from human, monkey, dog, mouse, rat, bovine, chicken, fish and Xenopus based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only