Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCOLCE Rabbit anti-Human, Mouse, Porcine, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA125190

Catalog No. PIPA125190

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-25190 detects PCOLCE from human, mouse, and porcine samples. PA1-25190 has been successfully used in Western blot procedures. The PA1-25190 immunogen is a synthetic peptide corresponding to the end of CUB2 and the beginning of the “linker” region of human PCOLCE. For coninuous use, store at 4°C for up to six months. For extended storage, the solution may be stored at -20°C. Do not store below -22°C.

Fibrillar collagen types I-III are synthesized as precursor molecules known as procollagens. These precursors contain amino- and carboxyl-terminal peptide extensions known as N- and C-propeptides, respectively, which are cleaved, upon secretion of procollagen from the cell, to yield the mature triple helical, highly structured fibrils. This gene encodes a glycoprotein which binds and drives the enzymatic cleavage of type I procollagen and heightens C-proteinase activity.
Specifications

Specifications

PCOLCE
Polyclonal
Unconjugated
PCOLCE
Liquid
PCOLCE
Synthetic peptide corresponding to the end of CUB2 and the beginning of the “linker” region of human PCOLCE.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Porcine
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.05% sodium azide
Q15113, Q61398
PCPE, PCPE1
Rabbit
IgG
25 μg
4° C, do not freeze
Primary
100627524, 18542, 5118
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit