Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Properdin, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA127525

Catalog No. PIPA127525

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Component of the ESCRT-III complex. which is required for multivesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. The MVB pathway mediates delivery of transmembrane proteins into the lumen of the lysosome for degradation.

PA1-27525 detects PCOLN3 from human samples.PA1-27525 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27501 immunogen is a synthetic peptide corresponding to human Charged multivesicular body protein 1a.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PCOLN3
Polyclonal
CFP
CHMP1, KIAA0047, PCOLN3, PRSM1, Vps46A
Chicken
Antigen affinity chromatography
RUO
5199
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Western Blot
Unconjugated
P27918
CFP
Synthetic peptide corresponding to human Charged multivesicular body protein 1a.
50 μg
Primary
Human
Antibody
IgY
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit