Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCSK2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514596

Catalog No. PIPA514596

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse, porcine and rat based on sequence homology.

PCSK2 belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This protein is a proinsulin-processing enzyme that plays a key role in regulating insulin biosynthesis. The protein is also known to cleave proopiomelanocortin, proenkephalin, prodynorphin and proluteinizing-hormone-releasing hormone.
Specifications

Specifications

PCSK2
Polyclonal
PBS with 0.09% sodium azide
P16519
Neuroendocrine convertase 2; NEC 2; Prohormone convertase 2; Proprotein convertase 2; PC2; KEX2-like endoprotease 2; NEC2; PCSK2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5126
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PCSK2
Liquid
PCSK2
KLH conjugated synthetic peptide between 318-346 amino acids from the central region of human PCSK2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit