Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCSK4, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89201713 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-201-713 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-201-713 Supplier Abnova Corporation Supplier No. H00054760K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human PCSK4 peptide using ARM Technology.

Proprotein convertases, including PCSK4, are calcium-dependent serine proteases related to bacterial subtilisins and to yeast kexin. These enzymes process precursor proteins to their active forms by selective cleavage of the polypeptide at sites following paired basic amino acids. In mammals, this family comprises PC1 (MIM 162150), PC2 (MIM 162151), PC4, PC5 (MIM 600488), furin (FUR; MIM 136950), and PACE4 (MIM 167405). Substrates for these enzymes range from prohormones to precursors for growth factors to cell surface receptors and viral surface glycoproteins (Cao et al., 2001 [PubMed 11776387]).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen PCSK4
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description proprotein convertase subtilisin/kexin type 4
Formulation In 1X PBS, pH 7.4
Gene PCSK4
Gene Accession No. PCSK4
Gene Alias DKFZp434B217, MGC34749, PC4, SPC5
Gene Symbols PCSK4
Host Species Rabbit
Immunogen A synthetic peptide of human PCSK4
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54760
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.