Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PCTAIRE1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232254

Catalog No. NBP232254


Add to cart

Description

Description

PCTAIRE1 Polyclonal antibody specifically detects PCTAIRE1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

PCTAIRE1
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
NP_006192.1
Cell division protein kinase 16, cyclin-dependent kinase 16, EC 2.7.11, EC 2.7.11.22, FLJ16665, PCTAIRE, PCTAIRE protein kinase 1, PCTAIRE1, PCTAIRE-motif protein kinase 1, PCTGAIRE, PCTK1Serine/threonine-protein kinase PCTAIRE-1
Rabbit
IgG
100 ul
Cell Cycle and Replication
Primary
5127
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
CDK16
The immunogen this antibody was made to, maps to a region between residue 25 to 75 of human PCTAIRE-Motif Protein Kinase 1 using the numbering given in entry NP_006192.1 (GeneID 5127).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only