Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCTAIRE3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP133487
Description
PCTAIRE3 Polyclonal specifically detects PCTAIRE3 in Human samples. It is validated for Western Blot.Specifications
PCTAIRE3 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
Cell division protein kinase 18, cyclin-dependent kinase 18, EC 2.7.11, EC 2.7.11.22, PCTAIRE, PCTAIRE3, PCTAIRE-motif protein kinase 3, PCTK3PCTAIRE protein kinase 3, Serine/threonine-protein kinase PCTAIRE-3 | |
Rabbit | |
54 kDa | |
0.1 mL | |
Protein Kinase | |
5129 | |
Human | |
IgG |
Western Blot | |
0.86mg/mL | |
Western Blot 1:500-1:3000 | |
Q07002 | |
CDK18 | |
Recombinant protein encompassing a sequence within the center region of human PCTAIRE3. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction