Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phosphate cytidylyltransferase 2, ethanolamine, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant PCYT2.

Manufacturer:  Abnova Corporation H00005833A01

Catalog No. 89-004-276


  Check Availability
Add to cart

Specifications

Description

Sequence: VWCDGCYDMVHYGHSNQLRQARAMGDYLIVGVHTDEEIAKHKGPPVFTQEERYKMVQAIKWVDEVVPAAPYVTTLETLDKYNCDFCVHGNDITLTVDGR
Description & Specifications

Specifications

phosphate cytidylyltransferase 2, ethanolamine
Unlabeled
phosphate cytidylyltransferase 2, ethanolamine
50% glycerol
NM_002861
PCYT2
PCYT2 (NP_002852, 25 a.a. ∽124 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5833
Western Blot
Human
Serum
PCYT2
ET
Murine
50μL
Human
Y
Polyclonal
Human
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit