Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCYOX1L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525523

Catalog No. PIPA525523

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Probable oxidoreductase.
Specifications

Specifications

PCYOX1L
Polyclonal
PBS with 0.09% sodium azide
Q8NBM8, Q8C7K6
prenylcysteine oxidase-like
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
240334, 78991
Immunohistochemistry, Western Blot
Unconjugated
PCYOX1L
Liquid
PCYOX1L
KLH conjugated synthetic peptide between 465-494 amino acids from the C-terminal region of human PCYXL
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit