Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

programmed cell death 11, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant PDCD11.

Manufacturer:  Abnova Corporation H00022984A01

Catalog No. 89-004-552


Add to cart

Description

Description

PDCD11 is a NF-kappa-B (NFKB1; 164011)-binding protein that colocalizes with U3 RNA (MIM 180710) in the nucleolus and is required for rRNA maturation and generation of 18S rRNA (Sweet et al., 2003 [PubMed 14624448]; Sweet et al., 2008 [PubMed 17654514]).[supplied by OMIM

Sequence: MANLEESFPRGGTRKIHKPEKAFQQSVEQDNLFDISTEEGSTKRKKSQKGPAKTKKLKIEKRESSKSAREKFEILSVESLCEGMRILGCVKEVNELELVISLPNGLQGFV
Specifications

Specifications

programmed cell death 11
Unconjugated
50% glycerol
BC049838
ALG-4|ALG4|KIAA0185|NFBP
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
22984
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant PDCD11.
PDCD11
Serum
PDCD11
PDCD11 (AAH49838, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit