Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDCD4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130866

Catalog No. PIPA130866


Add to cart

Description

Description

PA1-30866 detects PDCD4 from human samples. PA1-30866 has been successfully used in Western blot applications. The PA1-30866 immunogen is: Synthetic peptide derived from a region between amino acids 1-50 of Human PCDC4 (SwissProt ID Q99834). Note: The exact sequence is proprietary. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a protein localized to the nucleus in proliferating cells. Expression of this gene is modulated by cytokines in natural killer and T cells. The gene product is thought to play a role in apoptosis but the specific role has not yet been determined. Two transcripts encoding different isoforms have been identified.
Specifications

Specifications

PDCD4
Polyclonal
Unconjugated
PDCD4
Liquid
PDCD4
Synthetic peptide derived from a region between amino acids 1-50 of Human PCDC4 (SwissProt ID Q99834). Note: The exact sequence is proprietary.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q53EL6
RP11-348N5.4, H731, MGC33046, MGC33047
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
27250
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit