Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDCD5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526638
Description
This antibody is predicted to react with bovine based on sequence homology.
PDCD5 is a protein expressed in tumor cells during apoptosis independent of the apoptosis-inducing stimuli. Prior to apoptosis induction, this protein is distributed in both the nucleus and cytoplasm. Once apoptosis is induced, the level of this protein increases and by relocation from the cytoplasm, it accumulates in the nucleus. Although its exact function is not defined, this protein is thought to play an early and universal role in apoptosis.Specifications
PDCD5 | |
Polyclonal | |
PDCD5 | |
TFAR19, TF-1 cell apoptosis-related protein 19, TF1 cell apoptosis-related gene 19, TFAR19 novel apoptosis-related, programmed cell death protein 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56330, 9141 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
O14737, P56812 | |
PDCD5 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PDCD5 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction