Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE10A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against PDE10A.
Supplier: Abnova Corporation PAB12362
Description
Various cellular responses are regulated by the second messengers cAMP and cGMP. Phosphodiesterases, such as PDE10A, eliminate cAMP- and cGMP-mediated intracellular signaling by hydrolyzing the cyclic nucleotide to the corresponding nucleoside 5-prime monophosphate (Fujishige et al., 2000 [PubMed 10998054]).[supplied by OMIM
Specifications
PDE10A | |
Polyclonal | |
Rabbit polyclonal antibody raised against PDE10A. | |
In serum | |
FLJ11894/FLJ25677/HSPDE10A | |
Rabbit | |
50 μL | |
Primary | |
This antibody selectively detects proteins corresponding to the PDE10A. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (1:20000) The optimal working dilution should be determined by the end user. | |
PDE10A | |
PDE10A | |
C terminal region of human PDE10A. | |
RUO | |
10846 | |
Human, Mouse, Rat | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction