Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE11A Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131154

Catalog No. PA131154


Add to cart

Description

Description

PA1-31154 detects PDE11A4 from rat, mouse, human samples. PA1-31154 has been successfully used in Western blot and ELISA applications. The PA1-31154 immunogen is: Synthetic peptide (Rat) conjugated to KLH (N terminal).

Specifications

Specifications

PDE11A
Polyclonal
Unconjugated
PDE11A
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
140928, 241489, 50940
ELISA, Western Blot
0.75 mg/mL
proprietary buffer with 0.02% thimerosal
Q9HCR9, P0C1Q2, Q8VID6
PDE11A
Synthetic peptide (Rat) conjugated to KLH (N terminal).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit