Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE12 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523793

Catalog No. PIPA523793


Add to cart

Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

Specifications

Specifications

PDE12
Polyclonal
PBS with 0.09% sodium azide
Q6L8Q7
2′-PDE, 2′,5′-phosphodiesterase 12, 2′-phosphodiesterase, 2-PDE
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
201626
Flow Cytometry, Western Blot
Unconjugated
PDE12
Liquid
PDE12
KLH conjugated synthetic peptide between 362-390 amino acids from the C-terminal region of human PDE12
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit