Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

PDE1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131124

Catalog No. PA131124



Description

Description

PA1-31124 detects PDE1A from human, rat, mouse samples. PA1-31124 has been successfully used in immunoprecipitation, Western blot applications. The PA1-31124 immunogen is: Synthetic peptides (conserved sequence near the N-terminal) unique to PDE1A family members.

Cyclic nucleotide phosphodiesterases (PDEs) play a role in signal transduction by regulating intracellular cyclic nucleotide concentrations through hydrolysis of cAMP and/or cGMP to their respective nucleoside 5-prime monophosphates. Members of the PDE1 family, such as PDE1A, are Ca(2+)/calmodulin (see CALM1; MIM 114180)-dependent PDEs (CaM-PDEs) that are activated by calmodulin in the presence of Ca(2+) (Michibata et al., 2001 [PubMed 11342109]; Fidock et al., 2002 [PubMed 11747989]).
Specifications

Specifications

PDE1A
Polyclonal
Unconjugated
PDE1A
Liquid
PDE1A
Synthetic peptides (conserved sequence near the N-terminal) unique to PDE1A family members.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunoprecipitation, Western Blot
0.65 mg/mL
Antibody with no preservative
P54750, Q61481
HCAM1, HSPDE1A, MGC26303
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18573, 5136, 81529
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit