Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE4A Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131151

Catalog No. PA131151


Add to cart

Description

Description

PA1-31151 detects PDE4 from mouse, rat, human samples. PA1-31151 has been successfully used in Western blot applications. The PA1-31151 immunogen is: Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) C terminal. Store at 4°C short term. For extended storage aliquot and store at -20°C or below. Avoid freeze-thaw cycles.

Cyclic nucleotides are important second messengers that regulate and mediate a number of cellular responses to extracellular signals, such as hormones, light, and neurotransmitters. Cyclic nucleotide phosphodiesterases (PDEs) regulate the cellular concentrations of cyclic nucleotides and thereby play a role in signal transduction. PDE4A is a class IV cAMP-specific PDE (summary by Milatovich et al., 1994 [PubMed 8009369]).
Specifications

Specifications

PDE4A
Polyclonal
Unconjugated
PDE4A
Liquid
PDE4A
Synthetic peptide corresponding to the C terminal region.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
0.68 mg/mL
antibody with 0.02% thimerosal
P27815, O89084, P54748
DPDE2, PDE4, PDE46
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18577, 25638, 5141
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit