Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP133637
Description
PDE4B Polyclonal specifically detects PDE4B in Human samples. It is validated for Western Blot.Specifications
PDE4B | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal | |
PDE4B | |
Recombinant protein encompassing a sequence within the C-terminus region of human PDE4B. The exact sequence is proprietary. | |
0.1 mL | |
Cancer, Signal Transduction | |
5142 | |
Human | |
IgG |
Western Blot | |
0.72mg/mL | |
Western Blot 1:500-1:3000 | |
cAMP-specific (phosphodiesterase E4 dunce homolog, cAMP-specific 3'-5'-cyclic phosphodiesterase 4B, cAMP-specific phosphodiesterase-4 B isoform, DKFZp686F2182, Drosophila), dunce-like phosphodiesterase E4, EC 3.1.4, EC 3.1.4.17, MGC126529, PDE32, PDEIVB, phosphodiesterase 4B, cAMP-specific, phosphodiesterase 4B, cAMP-specific (dunce, phosphodiesterase 4B, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E4), phosphodiesterase E4 dunce homolog | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction