Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE4D Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131149

Catalog No. PA131149


Add to cart

Description

Description

PA1-31149 detects PDE4D5 from human, rat, mouse samples. PA1-31149 has been successfully used in Western blot applications. The PA1-31149 immunogen is: Synthetic peptide (unfortunately, the amino acid sequence is considered to be commercially sensitive) (N terminal).

Specifications

Specifications

PDE4D
Polyclonal
Unconjugated
PDE4D
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
238871, 24627, 5144
Western Blot
0.65 mg/mL
antibody with 0.02% sodium azide
P14270, Q08499, Q01063
PDE4D
Synthetic peptide corresponding to the N terminal region.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit