Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE6 gamma Rabbit anti-Bovine, Ovine, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1723

Catalog No. PA1723


Add to cart

Description

Description

PA1-723 detects phosphodiesterase 6 gamma (PDE6 gamma) from sheep retinal extract and bovine rod outer segment membranes as well as purified bovine PDE6 gamma. PA1-723 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~10 kDa protein representing PDE6 gamma from sheep retinal extract. The PA1-723 immunizing peptide corresponds to amino acid residues 1-15 from bovine PDE6 gamma. This peptide (Cat. No. PEP-150) is available for use in neutralization and control experiments.

The second messengers cAMP and cGMP are key regulatory molecules that are involved in a wide variety of signal transduction pathways. Levels of cAMP and cGMP are regulated by their rate of synthesis by nucleotide cyclases and by their rate of hydrolysis by cyclic nucleotide phosphodiesterases (PDEs). PDEs form a superfamily of enzymes that catalyze the conversion of 3-prime, 5-prime-cyclic nucleotides to the corresponding nucleoside 5-prime-monophosphates. PDE6 is the effector enzyme in the G protein-mediated signal transduction cascade in the visual system. There are five different subunits consisting of rod and cone specific catalytic subunits: alpha’ (Cone), alpha (Rod), and beta (Rod), the inhibitory subunit gamma, and subunit delta of unknown function (which likely interacts with many other proteins besides the PDE6 family). The catalytic core of the PDE6 system is comprised of alpha’/alpha’ homodimers in the cone and alpha/beta heterodimers in the rod. The C-terminus of both the catalytic and inhibitory subunits is modified by methylation, myristyolation and prenylation which have been shown to be critical for proper complex assembly and membrane association.
Specifications

Specifications

PDE6 gamma
Polyclonal
PBS with 1mg/mL BSA and 0.05% sodium azide
P04972
Phosphodiesterase 6 Gamma
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
101117520, 281977
Western Blot
Unconjugated
PDE6G
Liquid
PDE6G
Synthetic Peptide: M(1) N L E P P K A E I R S A T R(15) C
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Bovine, Ovine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit