Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE7B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524169

Catalog No. PIPA524169

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The 3 ft.,5 ft.-cyclic nucleotides cAMP and cGMP function as second messengers in a wide variety of signal transduction pathways. 3 ft.,5 ft.-cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and cGMP to the corresponding 5 ft.-monophosphates and provide a mechanism to downregulate cAMP and cGMP signaling. This gene encodes a cAMP-specific phosphodiesterase, a member of the cyclic nucleotide phosphodiesterase family.
Specifications

Specifications

PDE7B
Polyclonal
PBS with 0.09% sodium azide
Q9NP56
bA472E5.1, cAMP-specific 3′,5′-cyclic phosphodiesterase 7B, high-affinity cAMP-specific 3′,5′-cyclic phosphodiesterase, rolipram-insensitive phosphodiesterase type 7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27115
Western Blot
Unconjugated
PDE7B
Liquid
PDE7B
KLH conjugated synthetic peptide between 421-450 amino acids from the C-terminal region of human PDE7B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit