Learn More
MilliporeSigma™ anti-PDGF (Ab-1) (101-116), Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ PC213L100UL
Catalog No. PC213L100UL
Add to cart
Description
Description
Specifically detects PDGF (101-116) Clone: in Human, Rat samples, and it is validated for ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immuno Blotting
Platelet-Derived Growth Factor (PDGF) is the major serum mitogen for cells of mesenchymal origin in humans. The biologically active protein is a dimer composed of two related polypeptides designated A and B. The protein may exist as either a heterodimer AB or as a homodimer AA or BB. PDGF (BB) has been identified as the human homolog of the v-sis oncogene product and as such has been designated c-sis. The v-sis transforming domain is 89 amino acids in length and is identical in sequence to PDGF (BB). Binding of the PDGF molecule to its specific receptor occurs via interaction with a specific domain on the surface of the molecule known as the V4 surface domain. The PDGF protein has been implicated both directly as well as indirectly in several pathological states including neoplasia, arthritis, arteriosclerosis and bone marrow sclerosis.
Specifications
Specifications
PDGF (101-116)
Unconjugated
purified
a synthetic peptide (TEVFEISRRLIDRTNA) corresponding amino acids 101-116 of human PDGF B chain
100μL
2°C to 8°C
Polyclonal
Human, Rat
ELISA, Immuno Blotting, Immunoblot, Immunofluorescence, Immunohistochemistry (Paraffin)
In 0.05M sodium phosphate buffer, 0.2% gelatin.
Rabbit
IgG
RUO
Primary
Recognizes the B subunit of PDGF in mesenchymal cells and fetal epithelial tissue.