Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF-B Rabbit anti-Bovine, Canine, Feline, Hamster, Human, Mouse, Ovine, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA127394

Catalog No. PA127394

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-27396 detects Collagen I from mouse samples. PA1-27396 will not react with human, rat, or chicken. PA1-27396 has been successfully used in radioimmune assay, ELISA, immunofluorescence and immunocytochemistry procedures. This product is unsuitable for

Specifications

Specifications

PDGF-B
Polyclonal
PBS with 0.1% sodium azide; pH 7.2
Q6Q7I7, P12919, P01127, P31240, Q95229, Q05028
PDGFB
Synthetic peptide corresponding to residues T(101) E V F E I S R R L I D R T N A(116) of human PDGF beta.
IgG fraction
RUO
Antibody
Polyclonal
Bovine, Canine, Feline, Hamster, Human, Mouse, Ovine, Rat
Immunohistochemistry (Paraffin)
Unconjugated
PDGFB
Liquid
Rabbit
IgG
200 μL
4° C, do not freeze
Primary
100135774, 100774990, 18591, 24628, 442986, 443545, 5155, 540106
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit