Learn More
PDGF Goat anti-Bovine, Human, Mouse, Pig, Polyclonal, MilliporeSigma™
Goat Polyclonal Antibody
Manufacturer:  MilliporeSigma 06127
Catalog No. 06-127-MI
Add to cart
Description
Description
Anti-PDGF Antibody, neutralizing detects level of PDGF and has been published and validated for use in western blotting.
Specifications
Specifications
PDGF
Unconjugated
PDGFA; PDGF-1; PDGF-A; PDGF1
AB and BB.
DEAE chromatography
Primary
NM_033023, NM_033016, NM_016205
Western Blot
Purified
Goat
IgG
1mg
Polyclonal
Bovine, Human, Murine, Porcine