Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PDGF-RA/CD140a Clone: 7H13L1, Invitrogen™

PDGF-RA/CD140a Monoclonal antibody 701142 from Thermo Scientific™ Pierce™ specifically detects PDGF-RA/CD140a in Human samples, and it is validated for WB, IF, ICC and ChIP

Manufacturer:  Invitrogen 701142

Encompass

Catalog No. 70-114-2


Add to cart

Description

Description

PDGFR-alpha belongs to the protein tyrosine kinase family, and binds to the alpha chain of PDGF. Members of the PDGF receptor subfamily are cell surface receptors that act as mitogens for cells of mesenchymal origin, and consist of alpha and beta isoforms of ∼190, and 125 kDa, respectively. Activated PDGF receptors act on signaling pathways that control cell growth, actin reorganization, migration, and differentiation. In humans, the gene is located on the q arm of chromosome 4.

701142 detects PDGF-RA/CD140a in Human samples. 701142 has been successfully used in WB, IF, ICC and ChIP procedures. The immunogen for 701142 is recombinant protein corresponding to amino acids 259-436 of human platelet-derived growth factor receptor -

Applications:

Chromatin Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blotting

Specifications

Specifications

PDGF-RA/CD140a
Unconjugated
P16234
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Monoclonal
Human
7H13L1
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Purified
Recombinant protein corresponding to amino acids 259-436 of human platelet-derived growth factor receptor - alpha polypeptide
Protein A
Primary
5156
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit