Learn More
PDGFRA Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA517623
Catalog No. PIPA517623
Add to cart
Description
Description
It is not recommended to aliquot this antibody.
This gene encodes a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.
Specifications
Specifications
PDGFRA
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P26618, P16234
PDGF Receptor alpha, MGC74795, PDGFR2, RHEPDGFRA, CD140 antigen-like family member A, CD140a antigen, PDGF-R-alpha, PDGFRA/BCR fusion, pdgfr-a
Rabbit
IgG
100 μL
-20°C
Primary
18595, 5156
Flow Cytometry, Immunoprecipitation, Western Blot
Unconjugated
PDGFRA
Liquid
PDGFRA
Synthetic peptide corresponding to residues near the carboxy-terminal sequence of human PDGF receptor a
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse