Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFRB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138436

Catalog No. 01-673-842


Add to cart

Description

Description

This antibody is predicted to react with chicken, canine, amphibian, mouse, porcine, rabbit and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is prostate, skin or ovarian carcinoma.

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, alpha and beta coded by two different genes. PDGF-alpha receptors are expressed in oligodendrocyte progenitor cells and PDGF-beta receptors are expressed on neurons. This antibody is specific for PDGFR, beta and does not react with PDGFR, alpha.
Specifications

Specifications

PDGFRB
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P09619
PDGF Receptor beta, JTK12, PDGFR, PDGFR1, CD140 antigen-like family member B, PDGF-R-beta, beta-type platelet-derived growth factor receptor
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5159
Immunohistochemistry (Paraffin)
Unconjugated
PDGFRB
Liquid
PDGFRB
Synthetic peptide derived from near C-terminus of human PDGFR-beta
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit