Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PDGFRA/PDGFRB (Tyr572, Tyr574), Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128435

Catalog No. PA128435

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-28435 detects PDGFR alpha + beta from mouse, human samples.PA1-28435 has been successfully used in Western blot applications.The PA1-28435 immunogen is: Synthetic phospho peptide, containing Tyrosine 572 and 574 of Human PDGFR alpha. This corresponds to Tyrosine 579 and 581 of Human PDGFR beta. The sequence is conserved in PDGFR alpha mouse, rat and frog.
Specifications

Specifications

Phospho-PDGFR alpha + beta pTyr572 + pTyr574
Polyclonal
PBS with 50% glycerol, 1mg/mL BSA and 0.05% sodium azide; pH 7.3
P09619, P16234, P05622, P26618, P20786, Q05030, P26619
PDGF A-chain, PDGF subunit A, PDGF-1, platelet-derived growth factor A chain, platelet-derived growth factor alpha chain, platelet-derived growth factor alpha isoform 2 preproprotein, platelet-derived growth factor subunit A, pdgfr-a
Rabbit
IgG
50μL
Avoid Freeze/Thaw Cycles
Primary
5156, 5159
Western Blot
Unconjugated
PDGFRA
Purified
Pdgfra, pdgfra.L, Pdgfrb
Synthetic phospho peptide, containing Tyrosine 572 and 574 of Human PDGFR alpha. This corresponds to Tyrosine 579 and 581 of Human PDGFR beta. The sequence is conserved in PDGFR alpha mouse, rat and frog.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat, Xenopus laevis
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit