Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFR-Β Rabbit anti-Mouse, Human, Clone: 4A56, MilliporeSigma™

Rabbit Monoclonal Antibody

Manufacturer:  MilliporeSigma 05825R

Catalog No. 05-825-RMI

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Anti-PDGFR-β Antibody, clone 4A56 is an antibody against PDGFR-β for use in western blotting and immunoprecipitation.
Specifications

Specifications

PDGFR-β
4A56
Purified Rabbit Monoclonal IgG in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Purified
KLH-conjugated linear peptide corresponding to human PDGFR-β at the protein kinase domain.
Protein A
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Monoclonal
Human, Murine
Immunoprecipitation, Western Blot
Unconjugated
NP_002600
Rabbit
IgG
100μL
Primary
Becaplermin, PDGF-2, PDGF2, SIS, SSV, becaplermin, c-sis, PDGFRβ, PDGFR-β
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only