Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFRA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514709

Catalog No. PIPA514709

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PDGFRA is a cell surface tyrosine kinase receptor for members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.
Specifications

Specifications

PDGFRA
Polyclonal
PBS with 0.09% sodium azide
P16234
PDGF Receptor alpha, MGC74795, PDGFR2, RHEPDGFRA, CD140 antigen-like family member A, CD140a antigen, PDGF-R-alpha, PDGFRA/BCR fusion, pdgfr-a
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5156
Immunohistochemistry, Western Blot
Unconjugated
PDGFRA
Liquid
PDGFRA
KLH conjugated synthetic peptide between 740-769 amino acids from human PDGFRA
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit