Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
PDHA1 Mouse anti-Bovine, C. elegans, Drosophila, Human, Mouse, Rat, Zebrafish, Clone: 8D10E6, Invitrogen™ Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen 456600
Catalog No. 45-660-0
Description
Description
When performing ICC and IF heat-induced antigen-retrieval improves signal.|When performing Flow Cytometry Mouse monoclonal IgG1, is suitable for use as an isotype control with this antibody.|Positive Control: WB: Isolated mitochondria from human, bovine, rat and mouse heart. HepG2 (human liver hepatocellular carcinoma cell line) cell lysate. FACS: HeLa (human epithelial cell line from cervix adenocarcinoma) and HL-60 (human promyelocytic leukemia cell line) cells.
The pyruvate dehydrogenase (PDH) complex is a nuclear-encoded mitochondrial multienzyme complex that catalyzes the overall conversion of pyruvate to acetyl-CoA and CO(2), and provides the primary link between glycolysis and the tricarboxylic acid (TCA) cycle. The PDH complex is composed of multiple copies of three enzymatic components: pyruvate dehydrogenase (E1), dihydrolipoamide acetyltransferase (E2) and lipoamide dehydrogenase (E3). The E1 enzyme is a heterotetramer of two alpha and two beta subunits. This gene encodes the E1 alpha 1 subunit containing the E1 active site, and plays a key role in the function of the PDH complex.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
PDHA1
Monoclonal
1 mg/mL
HEPES buffered saline with 0.02% sodium azide
A7MB35, P08559, P35486, P26284
PDHA, PDHCE1A, PHE1A, PDHE1-A type I, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial, pyruvate dehydrogenase complex, E1-alpha polypeptide 1
Mouse
IgG1, kappa
100 μg
4° C, do not freeze
Primary
18597, 29554, 407109, 5160
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
8D10E6
Unconjugated
PDHA1
Liquid
PDHA1
Human recombit PDH-E1-alpha
Igg Fraction
RUO
Antibody
Monoclonal
Bovine, C. elegans, Drosophila, Human, Mouse, Rat, Zebrafish