Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDHA2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524247

Catalog No. PIPA524247

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The pyruvate dehydrogenase complex catalyzes the overall conversion of pyruvate to acetyl-CoA and CO(2). It contains multiple copies of three enzymatic components: pyruvate dehydrogenase (E1), dihydrolipoamide acetyltransferase (E2) and lipoamide dehydrogenase (E3).
Specifications

Specifications

PDHA2
Polyclonal
PBS with 0.09% sodium azide
P29803
PDHAL, PDHE1-A type II, pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, mitochondrial, pyruvate dehydrogenase, E1-alpha polypeptide, testis specific
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5161
Immunohistochemistry, Western Blot
Unconjugated
PDHA2
Liquid
PDHA2
KLH conjugated synthetic peptide between 287-314 amino acids from the central region of human PDHA2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit