Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDIA6, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127902

Catalog No. PIPA127902

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Protein disulfide isomerases (EC 5.3.4.1), such as PDIA6, are endoplasmic reticulum (ER) resident proteins that catalyze formation, reduction, and isomerization of disulfide bonds in proteins and are thought to play a role in folding of disulfide-bonded proteins (Hayano and Kikuchi, 1995 [PubMed 7590364]).

PA1-27902 detects PDIA6 from human, mouse, rat, and canine samples.PA1-27902 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Protein disulfide-isomerase A6.
Specifications

Specifications

PDIA6
Polyclonal
Unconjugated
PDIA6
Purified
PDIA6
Synthetic peptide corresponding to human Protein disulfide-isomerase A6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Human, Murine, Rat
Western Blot
1mg/mL
PBS with no preservative
Q15084, Q922R8, Q63081
ERP5, P5, TXNDC7
Chicken
IgY
50μg
Store at -20°C
Primary
10130
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit