Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyruvate Dehydrogenase Kinase 1/PDK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB1002383SS
Description
Pyruvate Dehydrogenase Kinase 1/PDK1 Polyclonal specifically detects Pyruvate Dehydrogenase Kinase 1/PDK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Pyruvate Dehydrogenase Kinase 1/PDK1 | |
Polyclonal | |
Unconjugated | |
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
EC 2.7.11, EC 2.7.11.2, mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1, PDH kinase 1, PDK1, PDK-1, PKB kinase, Pyruvate dehydrogenase kinase isoform 1, pyruvate dehydrogenase kinase, isoenzyme 1, pyruvate dehydrogenase kinase, isozyme 1, Pyruvate dehydrogenase lipoamide kinase isozyme 1, mitochondrial | |
Rabbit | |
49 kDa | |
0.025 mL | |
Cancer, Cytoskeleton Markers, Hypoxia, Lipid and Metabolism, Protein Kinase, Signal Transduction | |
5163 | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1.0 mg/ml | |
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:1000-1:2000 | |
Q15118 | |
PDK1 | |
A synthetic peptide made to a C-terminal portion of the human PDK1 protein sequence (between residues 350-436). [Swiss-Prot Q15118] | |
Affinity Purified | |
RUO | |
Primary | |
Human. Predicted to react with mouse and rat based on 100% sequence homology. Immunogen sequence has 93% homology to chicken. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction