Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyruvate Dehydrogenase Kinase 1/PDK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
$204.00 - $482.50
Specifications
Antigen | Pyruvate Dehydrogenase Kinase 1/PDK1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Immunoblotting reported in scientific literature (PMID 28366875) |
Applications | Western Blot, Immunoblot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1002384SS
|
Novus Biologicals
NB1002384SS |
0.025 mL |
Each for $204.00
|
|
|||||
NB1002384
|
Novus Biologicals
NB1002384 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
Pyruvate Dehydrogenase Kinase 1/PDK1 Polyclonal specifically detects Pyruvate Dehydrogenase Kinase 1/PDK1 in Human, Mouse samples. It is validated for Western Blot, Immunoblotting.Specifications
Pyruvate Dehydrogenase Kinase 1/PDK1 | |
Western Blot 0.5-2 ug/ml, Immunoblotting reported in scientific literature (PMID 28366875) | |
Polyclonal | |
Rabbit | |
Cancer, Cytoskeleton Markers, Hypoxia, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction | |
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
EC 2.7.11, EC 2.7.11.2, mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1, PDH kinase 1, PDK1, PDK-1, PKB kinase, Pyruvate dehydrogenase kinase isoform 1, pyruvate dehydrogenase kinase, isoenzyme 1, pyruvate dehydrogenase kinase, isozyme 1, Pyruvate dehydrogenase lipoamide kinase isozyme 1, mitochondrial | |
PDK1 | |
IgG | |
Affinity Purified | |
Human. Predicted to react with rat, mouse, primate and bovine based on 100% sequence homology. Immunogen sequence has 92% homology to chicken, 91% homology to Zebrafish and porcine. |
0.5 mg/mL | |
Western Blot, Immunoblot | |
Unconjugated | |
RUO | |
Human, Mouse | |
Q15118 | |
5163 | |
A synthetic peptide made to an internal portion of the human PDK1 protein sequence (between residues 300-400). [Swiss-Prot Q15118] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
49 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title