Learn More
anti-PDK-1, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP147237
Catalog No. NBP147237
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
PDK-1
0.5 mg/mL
Western Blot 1:1000
Purified
PDK1
Peptide corresponding to the amino acids CIQEVWRQRYQSHPDAAVQ of human PDK1
63 kDa
0.1 mg
Autophagy, Cancer, Cytoskeleton Markers, Hypoxia, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction
Primary
5163
Human
Western Blot
Unconjugated
PBS and 0.2% (w/v) high grade BSA (protease free) with 15mM Sodium Azide
[Pyruvate dehydrogenase [lipoamide]] kinase isozyme 1, mitochondrial, EC 2.7.11, EC 2.7.11.2, mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1, Pyruvate dehydrogenase kinase isoform 1, pyruvate dehydrogenase kinase, isoenzyme 1, pyruvate dehydrogenase kinase, isozyme 1
Rabbit
IgG
Protein A purified
RUO
Store at 4C. Do not freeze.
Polyclonal
The rabbit polyclonal antibody recognizes PDK1, a serine/threonine proteinkinase of 63 kDa, which is a central activator of multiple signaling pathways coupled to a large number of growth-promoting stimuli.